Learn More
Medchemexpress LLC MIP-1 alpha/CCL3 Mo 500ug | 500ug
SDP
Supplier:  Medchemexpress LLC HYP7768500UG
Encompass_Preferred
MIP-1 alpha/CCL3 Protein Mouse (His) an important chemokine is a key regulator of immune microenvironment and primarily mediates the trafficking of immune cells in both inflammation and cancer MIP-1 alpha/CCL3 Protein Mouse (His) is a recombinant mouse CCL3 (A24-A92) expressed by E coil with a N-6 His tag[1]
Catalog No. 50-001-87299