Learn More
Medchemexpress LLC MIP-3 beta/CCL19 Mo 10ug
SDP
Supplier:  Medchemexpress LLC HYP726410UG
Encompass_Preferred
MIP-3 beta/CCL19 Protein Mouse is a CC chemokine that is strongly chemotactic for CD4 T cells and CD8 T cells and acts as a ligand that binds specifically to the chemokine receptor CCR7 to mediate tissue immunity inflammatory responses and antiviral infections MIP-3 beta/CCL19 Protein Mouse is a recombinant mouse MIP-3 beta/CCL19 (G26-S108) protein expressed by E coli[1][2]
Catalog No. 50-001-82619