Learn More
Medchemexpress LLC MIP-4/CCL18 Human 5ug
SDP
Supplier:  Medchemexpress LLC HYP72655UG
Encompass_Preferred
MIP-4/CCL18 Protein Human is a CC chemokine ligand that binds to PITPNM3 GPR30 and CCR8 receptors and also acts as a neutral CCR3 antagonist mediating inflammation autoimmunity and carcinogenesis MIP-4/CCL18 Protein Human is a recombinant human MIP-4/CCL18 (A21-A89) protein expressed by E coli[1][2]
Catalog No. 50-001-82624