Learn More
Selleck Chemical LLC MIRA-1 E4634-5MG
SDP
Supplier:  Selleck Chemical LLC E46345MG
Encompass_Preferred
MIRA-1(NSC 19630) a maleimide compound is a selective inhibitor of Werner syndrome WRN helicase activity with an IC50 of 20 M while sparing ATPase and exonuclease activities It reactivates mutant p53 inducing apoptosis in various human solid tumor cell lines with tetracycline-regulated mutant p53
Catalog No. 50-000-39047