Learn More
Abcam MIRA-1, p53 inducer, 50MG
Supplier:  Abcam AB14586250MG
Encompass_Preferred
MW 183.16 Da, Purity >99%. Novel, nontoxic and selective p53 inducer. Antitumor agent. Promotes tumor cell apoptosis in a mutant p53-dependent manner (IC₅₀ = 10 μM) and prolongs survival. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5066