Learn More
Medchemexpress LLC Mirabegron | 223673-61-8 | MFCD11100356 | 99.9% | 396.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14773100MG
Encompass_Preferred
Mirabegron is a selective β3-adrenoceptor agonist, demonstrating potent bladder relaxant properties. It is an orally active compound known for its ability to increase cyclic AMP accumulation, primarily targeting β3-adrenoceptors with an EC50 of 22.4 nM. This compound exhibits significant selectivity, showing minimal agonistic effects on β1- and β2-adrenoceptors.
  • Selective β3-adrenoceptor agonist
  • Increases cyclic AMP accumulation
  • Potent bladder relaxant compound
  • Exhibits high selectivity over β1- and β2-adrenoceptors
  • Effective in treating overactive bladder symptoms such as urgency and frequency
Catalog No. 50-003-60742