Learn More
Selleck Chemical LLC Mirabegron 5mg 223673-61-8 YM 178
SDP
Supplier:  Selleck Chemical LLC S40095MG
Encompass_Preferred
Mirabegron (YM 178) is a selective ?3-adrenoceptor agonist with EC50 of 22.4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3528