Learn More
Apexbio Technology LLC Mirabegron (YM178), 10mg. Cas: 223673-61-8 MFCD: MFCD11100356. Selective β3-adrenoceptor agonist
SDP
Supplier:  Apexbio Technology LLC A847410
Encompass_Preferred
Mirabegron activates the 3 adrenergic receptor in the detrusor muscle in the bladder, which leads to muscle relaxation and an increase in bladder capacity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5149