Learn More
Selleck Chemical LLC Miransertib (ARQ 092) HCl 5mg 1313883-00-9 Miransertib
SDP
Supplier:  Selleck Chemical LLC S83395MG
Encompass_Preferred
Miransertib (ARQ 092) HCl is a novel, orally bioavailable and selective AKT pathway inhibitor exhibiting a manageable safety profile among patients with advanced solid tumors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5966