Learn More
Enzo Life Sciences Mirin (25mg). CAS: 299953-00-7
SDP
Supplier:  Enzo Life Sciences BMLGR3570025
Encompass_Preferred
The MRN-ATM pathway is essential for sensing the signaling from DNA double-strand breaks. Mirin is an inhibitor of MRN discovered using a forward chemical genetic screen. It prevents MRN-dependent activation of ATM without affecting ATM protein kinase activity and it inhibits Mre11-associated exonuclease activity. IC50=66μM. It abolishes the G2/M checkpoint and homology-dependent repair in mammalian cells. Alternative name: Z-5-(4-Hydroxybenzylidene)-2-imino-1,3-thiazolidin-4-one. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25mg/ml) or DMF (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1005