Learn More
Medchemexpress LLC Mirodenafil | 862189-95-5 | 99.9% | 531.67 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14930100MG
Encompass_Preferred
Mirodenafil (SK3530) is an orally active, potent, reversible, and selective phosphodiesterase 5 (PDE5) inhibitor. It also acts as a glucocorticoid receptor (GR) modulator and activates the Wnt/β-catenin signaling pathway by downregulating Dkk1 expression. Mirodenafil is used in research for erectile dysfunction (ED), Alzheimer's disease (AD), and systemic sclerosis (SSc).
  • Orally active
  • Potent, reversible, and selective PDE5 inhibitor
  • Glucocorticoid receptor (GR) modulator
  • Activates Wnt/β-catenin signaling pathway by downregulating Dkk1 expression
  • Enhances neuronal survival by protecting mitochondrial membrane potential and inhibiting apoptosis
  • Inhibits GSK-3β signaling, reducing tau phosphorylation and Aβ production
  • Enhances cognitive-behavioral performance in transgenic AD mice
Catalog No. 50-003-60973