Learn More
Enzo Life Sciences Misoprostol (100mg). CAS: 59122-46-2
SDP
Supplier:  Enzo Life Sciences BMLPG0510100
Encompass_Preferred
A cytoprotective PGE1 analog. Prevents NSAID-induced gastric ulceration. Misoprostol is a methyl ester and is therefore inactive in receptor preparations and in other situations where it does not undergo ester hydrolysis. Alternative name: SC-29333. Purity: ≥99% (TLC). Solubility: Soluble in ethanol (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1427