Learn More
Cayman Chemical Misoprostol free acidd5
SDP
Supplier:  Cayman Chemical 10010333100 UG
Encompass_Preferred
An internal standard for the quantification of misoprostol (free acid) by GC- or LC-MS.
Catalog No. 50-289-1136