Learn More
ATCC MITC-STO; MITC-Treated Fibroblast; Mouse (Mus musculus)
Supplier:  ATCC 56X.2
Encompass
MITC-STO mitomycin C-treated feeder cells are fibroblasts derived from a mouse embryo. These cells can be used as feeder cells to support the growth of other cells. This cell line has applications in stem cell research.
Catalog No. 50-238-1803