Learn More
Medchemexpress LLC Mitiglinide calcium hydrate | 207844-01-7 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0682A10MM/1ML
Encompass_Preferred
Mitiglinide calcium hydrate is an insulinotropic agent and an ATP-sensitive K+ (KATP) channel antagonist, exhibiting high specificity for the pancreatic beta-cell KATP channel (Kir6.2/SUR1 complex). It is utilized in research for type 2 diabetes.
  • Acts as an insulin secretagogue
  • Specific KATP channel antagonist
  • Potently inhibits Kir6.2/SUR1 channel currents
  • Reduces post-meal blood glucose levels
  • Useful in diabetes research
Catalog No. 50-004-07713