Learn More
Medchemexpress LLC Mito-DK | 95.1% | 608.49 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16242250MG
Encompass_Preferred
Mito-DK is a small-molecule fluorescent dye that offers crosstalk-free response to polarity and mtDNA, as well as mitochondrial morphology. It exhibits high photostability, low cytotoxicity, and good mitochondria-targeting properties. It can be used for real-time tracking and multidimensional assessment of mitochondria-related pyroptosis in cancer cells.
  • Small-molecule fluorescent dye
  • Crosstalk-free response to polarity and mtDNA
  • Crosstalk-free response to mitochondrial morphology
  • High photostability
  • Low cytotoxicity
  • Good mitochondria-targeting properties
  • Can be used for real-time tracking and multidimensional assessing of mitochondria-related pyroptosis in cancer cells
Catalog No. 50-003-92872