Learn More
Selleck Chemical LLC Mito-TEMPO 5mg
SDP
Supplier:  Selleck Chemical LLC S97335MG
Encompass_Preferred
Mito-TEMPO (MT), a mitochondria-targeted superoxide dismutase mimetic, protects against the early phase of acetaminophen (APAP) hepatotoxicity by inhibiting peroxynitrite formation. Mito-TEMPO treatment inhibits APAP-inducedRIP3 kinase expression. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3506