Learn More
Selleck Chemical LLC Mito-TEMPO S9733-25MG
SDP
Supplier:  Selleck Chemical LLC S973325MG
Encompass_Preferred
Mito-TEMPO (MT) a mitochondria-targeted superoxide dismutase mimetic protects against the early phase of acetaminophen (APAP) hepatotoxicity by inhibiting peroxynitrite formation Mito-TEMPO treatment inhibits APAP-induced RIP3 kinase expression
Catalog No. 50-000-24908