Learn More
Medchemexpress LLC MitoBloCK-10 5mg | 394694-98-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1154675MG
Encompass_Preferred
MitoBloCK-10 (MB-10) is a small-molecule Tim44 inhibitor that modulates the protein-associated motor (PAM) complex and is used in research to study mitochondrial protein import and related pathways.
  • Inhibits Tim44 and attenuates PAM complex activity.
  • Useful for studying mitochondrial protein import and quality control.
  • Supplied as a solid research-grade compound, typically in a 5 mg pack.
  • Molecular formula C12H8FN3O3S and molecular weight 293.27.
Catalog No. 50-002-28903