Learn More
Cayman Chemical MitoCheck Complex II Activity Assay Kit, 96 Well
SDP
Catalog No. 502892348
Encompass_Preferred
Complex II (succinate dehydrogenase/co-enzyme Q reductase) is one of the major sites of electron entry into the mitochondrial electron transport chain (ETC). Complex II catalyzes the oxidation of succinate to fumarate, and in the process reduces ubiquinone (Q) to ubiquinol (QH2). Ultimately, oxidation of succinate will lead to reduction of O2, the terminal step in mitochondrial respiration. Cayman’s MitoCheck Complex II Activity Assay allows for the activity of complex II to be determined in isolated mitochondria. As complex II oxidizes succinate, electrons are passed to an analog of ubiquinone and then on to DCPIP, which, when oxidized, absorbs in the 600 nm range. The absorbance of DCPIP will decrease upon reduction. Thus, Complex II activity is measured as a decrease in absorbance at 600 nm over time.
Catalog No. 50-289-2348 Supplier Cayman Chemical Supplier No. 70094096 WELL
Complex II (succinate dehydrogenase/co-enzyme Q reductase) is one of the major sites of electron entry into the mitochondrial electron transport chain (ETC). Complex II catalyzes the oxidation of succinate to fumarate, and in the process reduces ubiquinone (Q) to ubiquinol (QH2). Ultimately, oxidation of succinate will lead to reduction of O2, the terminal step in mitochondrial respiration. Cayman’s MitoCheck Complex II Activity Assay allows for the activity of complex II to be determined in isolated mitochondria. As complex II oxidizes succinate, electrons are passed to an analog of ubiquinone and then on to DCPIP, which, when oxidized, absorbs in the 600 nm range. The absorbance of DCPIP will decrease upon reduction. Thus, Complex II activity is measured as a decrease in absorbance at 600 nm over time.