Learn More
Sigma Aldrich Fine Chemicals Biosciences Mitochondria/Cytosol fract
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MIT1000
Encompass
Mitochondria sometimes described as the power plants of the cell are sites where most of the energy production in eukaryotic cells takes place. The synthesis of most of the adenosine triphosphate (ATP) occurs in these double membraned organelles that are found in living cells. ATP production by the mitochondria is done by the process of respiration which uses oxygen to generate energy. This is a very efficient process for using food energy to make ATP. In addition to supplying cellular energy mitochondria are involved in a range of other processes such as signaling cellular differentiation cell death as well as the control of the cell cycle and cell growth. Mitochondria also regulate crucial apoptosis signaling pathways. The number of mitochondria in a cell varies widely by organism and tissue type. Many cells have only a few mitochondria whereas others can contain several thousand.Millipores Mitochondria/Cytosol fractionation Kit provides reagents for quick and efficient isol
Catalog No. 50-290-4361