Learn More
Enzo Life Sciences Mitochondria/Cytosol fractionation kit (1 Kit)
SDP
Supplier:  Enzo Life Sciences ALX850276KI01
Encompass_Preferred
The core kit contains unique formulations of buffers and reagents that allow isolation of mitochondria and cytosol in only two easy steps. No ultracentrifugations are required. No toxic chemicals are involved. Species reactivity: Mammalian.
Catalog No. 50-201-0149