Learn More
Medchemexpress LLC Mitochondria degrader-1 | 2241669-05-4 | 95.1% | 758.30 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147108
Encompass_Preferred
Mitochondria degrader-1 is a potent compound engineered to induce the degradation of injured mitochondria through the autophagy mechanism. This degrader facilitates the removal of damaged mitochondria, making it a valuable tool for research in various disease areas.
  • Valuable tool for research in various disease areas
  • Applications span across studies related to neurodegenerative diseases, cancer, inflammatory conditions, age-related diseases, metabolic disorders, mitochondrial diseases, and Down's disease
Catalog No. 50-003-64002