Learn More
Biotium Primary Antibody Mitochondrial Marker 2509.MTC CF740 1/EA
Supplier:  Biotium P045740500UL
Encompass_Preferred
Mitochondrial Marker (2509 MTC) is a recombinant mouse monoclonal antibody that recognizes a 60 kDa antigen associated with the mitochondria in cells This CF740 antibody conjugate has been validated in IF CF and reg dyes are fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00464