Learn More
Sigma Aldrich Fine Chemicals Biosciences Gelatin SepharoseR 4B Cyti
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17095601
Encompass
Gelatin Sepharose(R) 4B affinity media with immobilized gelatin is designed for the purification or depletion of fibronectin as gelatin binds to fibronectin with high specificity. Gelatin Sepharose(R) 4B is bovine gelatin coupled to Sepharose(R) 4B by the cyanogen bromide method.
Catalog No. 50-290-4852