Learn More
TARGETMOL CHEMICALS INC MITOCHONIC ACID 5 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T836610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mitochonic acid 5 (MA-5) is a derivative of the plant hormone indole-3-acetic acid. Mitochonic acid 5 binds mitochondria and ameliorates renal tubular and cardiac myocyte damage.It has been shown to improve survival of fibroblasts from patients with mitochondrial diseases. purity: 99%
Catalog No. 50-257-9086