Learn More
Medchemexpress LLC Mitoguazone 25mg | 459-86-9 | 25MG
SDP
Supplier:  Medchemexpress LLC HY10663425MG
Encompass_Preferred
Mitoguazone (Methylglyoxal-bis(guanylhydrazone)) is a synthetic polycarbonyl derivative with potent antineoplastic activity Mitoguazone is a brain-penetrant and competitive S-adenosyl-methionine decarboxylase (SAMDC) inhibitor that disrupts polyamine biosynthesis Mitoguazone induces cell apoptosis Mitoguazone inhibits HIV DNA integration into the cellular DNA in both monocytes and macrophages Mitoguazone has the potential for acute leukemia Hodgkin s and non-Hodgkin s lymphoma treatment[1 [2 [3 [4
Catalog No. 50-002-42019