Learn More
Apexbio Technology LLC Mitomycin C, 50-07-7, MFCD00078109, 10 mg
SDP
Supplier:  Apexbio Technology LLC A445210
Encompass_Preferred
MF: C15H18N4O5, MW: 334.33, Solubility: 16.7mg/mL in DMSO,insoluble in EtOH,insoluble in H2O, Synonyms: Ametycine. Mitomycin C, a kind of antibiotic isolated from Streptomyces caespitosus or Streptomyces lavendulae, inhibits DNA synthesis through covalent mitomycin C-DNA adduct with EC50 values of 0.14M in PC3 cells. Therefore, it was served as a chemotherapeutic agent that has demonstrated its antitumor activity and has been used widely in treatment of various cancers.
Catalog No. 50-101-0599