Learn More
Selleck Chemical LLC Mitomycin C 50mg 50-07-7
SDP
Supplier:  Selleck Chemical LLC S814650MG
Encompass_Preferred
Mitomycin C is an antineoplastic antibiotic by inhibiting DNA synthesis, used to treat different cancers. Mitomycin C induces apoptosis in a caspases-dependent and Fas/CD95-independent manner. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5716