Learn More
Sigma Aldrich Fine Chemicals Biosciences Mitomycin C from Streptomyces caespitosus, 50-07-7, MFCD00078109, 2mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M42872MG
Encompass
Empirical Formula (Hill Notation) C15H18N4O5, Molecular Weight 334.33, powder, BioReagent, suitable for cell culture
Catalog No. 50-165-7011