Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MitoPQ, selective redox cycler, 5 mg, solid, >98% purity, 954.5 Da, C39H46I3N2P, shipping on blue ice, store at +4°C

Supplier:  Abcam AB1468195MG

Encompass_Preferred

MitoPQ is a selective and mitochondria-targeted redox cycler, specifically designed to enhance superoxide production in the mitochondrial matrix in both in vitro and in vivo conditions. This innovative compound is composed of a triphenylphosphonium lipophilic cation linked to the redox cycler paraquat, making it a vital tool for research into mitochondrial superoxide's role in pathology and redox signaling.

  • Selective, mitochondria-targeted redox cycler
  • Increases superoxide production in the mitochondrial matrix
  • Composed of triphenylphosphonium and paraquat
  • Molecular weight: 954.5 Da
  • Purity: >98%
  • CAS Number: 1821370-28-8
  • Form: Solid
  • Molecular formula: C39H46I3N2P
  • Solubility in ethanol: 10 mM
  • Solubility in DMSO: 100 mM
  • Shipping conditions: Blue Ice
  • Storage conditions: Store under desiccating conditions at +4°C

Catalog No. NC1226676


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.