Learn More
Medchemexpress LLC Mitoquinone mesylate | 845959-50-4 | 678.81 g·mol⁻¹ | C38H47O7PS | 1 ML

Supplier: Medchemexpress LLC HY100116A10MM 1ML
Mitoquinone mesylate is a triphenylphosphonium-conjugated ubiquinone antioxidant that selectively accumulates in mitochondria to reduce reactive oxygen species and protect against oxidative damage. Provided as a prepared 10 mM solution (1 mL vial) for research use, the product includes solubility and storage guidance for experimental handling.
- Mitochondria-targeted antioxidant mechanism reduces ROS.
- Triphenylphosphonium-conjugated ubiquinone structure for selective accumulation.
- Ready-to-use 10 mM solution in a 1 mL vial for convenience.
- Solubility in water, ethanol, and DMSO facilitates formulation.
- Storage recommendations for long-term stability included.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.