Learn More
Medchemexpress LLC Mitoquinone mesylate | 845959-50-4 | MFCD30747833 | 99.1% | 678.81 g·mol⁻1 | C38H47O7PS | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100116A100MG
Encompass_Preferred
Mitoquinone mesylate is a triphenylphosphonium (TPP)-based, mitochondrially targeted antioxidant used in research to protect against mitochondrial oxidative damage by scavenging reactive oxygen species (ROS). It is provided as a research-grade solid suitable for preparation of in vitro and in vivo test solutions for studies of oxidative stress and mitochondrial biology.
  • Targets mitochondrial reactive oxygen species (ROS).
  • Triphenylphosphonium (TPP)-based structure for mitochondrial delivery.
  • Suitable for oxidative stress and mitochondrial function studies.
  • Supplied as a solid reagent for flexible preparation and dosing.
  • High purity to support reproducible experimental results.
Catalog No. 50-002-15201