Learn More
Medchemexpress LLC HY-126222A 5mg , MitoTam (iodide, hydriodide) CAS:1634624-74-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY126222A5MG
Encompass_Preferred
HY-126222A 5mg MitoTam (iodide, hydriodide) CAS: 1634624-74-0 MitoTam iodide, hydriodide is a Tamoxifen derivative, an electron transport chain (ETC) inhibitor, spreduces mitochondrial membrane potential in senescent cells and affects mitochondrial morphology.MitoTam iodide, hydriodide is an effective anticancer agent, suppresses respiratory complexes (CI-respiration) and disrupts respiratory supercomplexes (SCs) formation in breast cancer cells. MitoTam iodide, hydriodide causes apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1723