Learn More
TARGETMOL CHEMICALS INC Mitotane 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T11991G
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Mitotane (NCI-C04933) is a derivative of the insecticide DICHLORODIPHENYLDICHLOROETHANE that specifically inhibits cells of the adrenal cortex and their production of hormones. It is used to treat adrenocortical tumors and causes CNS damage, but no bone marrow depression. Purity 99.92%
Catalog No. 50-287-5908