Learn More
TARGETMOL CHEMICALS INC Mitoxantrone dihydrochloride
SDP
Supplier:  TARGETMOL CHEMICALS INC T0158500MG
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mitoxantrone dihydrochloride (NSC-301739) is the hydrochloride salt of an anthracenedione antibiotic with antineoplastic activity. It is a type II topoisomerase inhibitor. Purity 98.63%
Catalog No. 50-288-0775