Learn More
Medchemexpress LLC HY-N6946 5mg Medchemexpress, Mitraphylline CAS:509-80-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69465MG
Encompass_Preferred
Medchemexpress, HY-N6946 5mg Mitraphylline CAS:509-80-8 Mitraphylline is the major pentacyclic oxindolic alkaloid presented in Uncaria tomentosa. Mitraphylline inhibits lipopolysaccharide-mediated activation of primary human neutrophils. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1591