Learn More
McMaster-Carr MITUTOYO 10-PIN TO USB ADAPTER
Supplier:  McMaster-Carr 1825A14
Encompass
Mitutoyo Adapter, Model 264-020, 10-Pin to USB-Adapt Mitutoyo 10-pin SPC cables for connection to a USB port on a PC to import measurement data into Microsoft Excel or any SPC software.
Catalog No. NC2295514