Learn More
MSC Mitutoyo Dial Bore Gage, 1.4 to 2.5 in, 0.000080 in Accuracy
Supplier:  MSC 75127936
Encompass
The Dial Bore Gage is designed for measuring internal diameters with high precision. It features a broad measurement range and high accuracy, making it suitable for demanding laboratory and industrial applications. This robust gage is also noted for its ease of use and durability.
  • Measures bore diameters from 1.4 to 2.5 inches.
  • Provides 0.000080 inch accuracy.
  • Measures ovalness and taper.
  • Features a two-point contact system for reliable readings.
  • Equipped with a pendulum guide structure.
  • Includes a measurement indicator for immediate feedback.
  • Comes with various accessories for versatile applications.
  • Designed for ease of setting and use, even in challenging conditions.
Catalog No. 50-005-31424