Learn More
MSC Mitutoyo Dial Caliper, 0-6 in, 0.001 in Graduation, Carbide-Tipped Jaws, White Face
Supplier: MSC 84477843
This dial caliper offers precise measurements for various applications. It features a 0-6 inch range, 0.001 inch graduation, and hardened stainless steel construction. Carbide-tipped jaws enable accurate outside diameter measurements, while the caliper handles depth, inside diameter, outside diameter, and step measurements. It is supplied in a fitted plastic case.
- Hardened stainless steel construction for durability.
- Smooth, high-shock protection dial movement.
- Measures OD, ID, depth, and steps.
- Carbide-tipped jaws for precise outside measurements.
- Thumb roller ensures consistent, accurate readings.
- TiN coated sliding surfaces reduce wear.
- Micro-fine graduations and adjustable bezel.
- Covered rack for clean gear teeth.
- Includes fitted plastic case and inspection certificate.