Learn More
MSC Mitutoyo Dial Caliper, 6 inch, 0.001 inch Graduation, Carbide-Tipped Jaws
Supplier: MSC 84477892
This dial caliper, constructed from hardened stainless steel, offers precise measurements for outside, inside, step, and depth dimensions. Its durable design ensures long-lasting performance in professional settings.
- New dial movement provides ultra-smooth sliding and high-shock protection.
- Improved finish on sliding surfaces ensures longevity and enhanced readability.
- Lock screw for dial bezel and sliding jaw position.
- Carbide-tipped jaws for accurate OD measurements.
- Supplied in a fitted plastic case for protection.
- Thumb roller allows for consistent feel and accurate measurements.
- Titanium nitride (TiN) coated main beam sliding surfaces reduce wear.
- Extra rigid main beam with micro-fine graduations.
- Adjustable and locking bezel.
- Covered rack keeps gear teeth clean for easy zero adjustment.