Learn More
Grainger Mitutoyo Digimatic Caliper, Absolute Encoder, 0 to 6 in., 0.0005 in. Resolution, Carbide Jaws
Supplier: Grainger 50017532
The Mitutoyo Digimatic Caliper is equipped with absolute encoder technology, allowing it to maintain its origin point once set. It offers a measuring range of 0 to 6 in. and a resolution of 0.0005 in. This caliper features carbide-tipped jaws, suitable for both inside and outside diameter measurements, and provides SPC data output for efficient data management.
- Absolute encoder technology keeps track of the origin point once set.
- SPC data output.
- Large and clear LCD readout.
- ZERO/ABS key allows zero-setting at any slider position for incremental comparison measurements and return to the absolute coordinate.
- Optional data hold unit.
- Carbide-tipped jaws for measurement of inside and outside diameter.