Learn More
MSC Mitutoyo Digimatic Holtest Internal Micrometer Set, 0.5-0.8 in, Three-Point, with Extension Rod
Supplier:  MSC 72958812
Encompass
This Digimatic Holtest Three-Point Internal Micrometer Set provides precise internal measurements with a range of 0.5 to 0.8 inches. It features a digital display for easy readability and includes an extension rod, making it suitable for measuring deep holes and to the bottom of blind holes. The micrometer is designed for consistent and accurate results in various applications.
  • Titanium-coated measuring pins provide excellent durability and impact resistance.
  • Measures accurately to the bottom of blind holes.
  • Large LCD display for clear readouts.
  • Equipped with a ratchet stop for constant measuring force.
  • Supports absolute and incremental measuring modes.
  • Features data hold, data output, and preset functions.
  • Inch/millimeter conversion available on inch/metric models.
  • Long battery life for extended use.
Catalog No. 50-005-27707