Learn More
MSC Mitutoyo Electronic Depth Gage, 0-12 in, 0.0005 in Resolution, ABSOLUTE Digimatic
Supplier:  MSC 14345698
Encompass
The Mitutoyo ABSOLUTE Digimatic Depth Gage provides highly accurate depth measurements for any shape that has a step, groove or slot. It features hardened, micro-lapped measuring faces and a base for improved accuracy and wear resistance. Constructed of hardened stainless steel, it keeps track of the origin point once set for the entire life of the battery.
  • Origin set function
  • Zero setting
  • Automatic power on/off
  • Inch/mm conversion
  • SPC output function with optional water-resistant SPC cable
  • Includes battery
  • Water-resistant with IP67 protection
  • Constructed of hardened stainless steel
  • Traceability certification included (NIST)
  • Calibrated
  • ABSOLUTE (ABS) linear scale eliminates the need for setting the origin point every time the micrometer is powered on
Catalog No. 50-005-29497