Learn More
MSC Mitutoyo Electronic Height Gage, 40 in, 0.0005 in Resolution, 0.003 in Accuracy
Supplier:  MSC 01484500
Encompass
This electronic height gage offers precise and reliable measurement for various applications. With a maximum measurement of 40 inches and a resolution of 0.0005 inch, it provides accurate readings. The gage features a large LCD for improved visibility and includes a scriber for marking.
  • ABS/INC switching
  • Data output
  • Zero setting
  • Two presets
  • Coarse/fine feeding switch
  • Data hold
  • Inch/mm conversion
  • Inclined slider handle
  • Large LCD characters
  • Power on/off
  • Large function buttons
  • Scriber included
  • Absolute origin setting
  • Incremental measurement
  • Smooth vertical movement with slider handle
  • Secure clamping with large lever
Catalog No. 50-005-29618