Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Mitutoyo Flat Head Screw, For Square Gage Blocks, 15.8 mm Length

Supplier:  MSC 03526282

Encompass

This flat head screw is designed for use with square gage blocks. It is inserted into the center hole of the square gage block to secure components. The screw measures 15.8 mm in length and has a diameter of 4.166 mm.

  • Designed for use with square gage blocks
  • Inserts into the center hole of the square gage block
  • Measures 15.8 mm in length
  • Has a diameter of 4.166 mm

Catalog No. 50-005-29639


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.