Learn More
MSC Mitutoyo Gage Block, Rectangle, 2 Inch, Steel, Grade 0
Supplier:  MSC 06178677
Encompass
This gage block is a precision instrument designed for accurate measurements. It features a rectangular shape, is made from durable steel, and has an accuracy grade of 0. This product is suitable for various metrology applications requiring high precision.
  • Rectangle shape
  • Accuracy grade of 0
  • Includes certificate of inspection
  • Made from steel
  • 2 in overall length
  • Complies with ASME standards
Catalog No. 50-005-29923