Learn More
MSC Mitutoyo Pocket Indicator Stem, 3/8 in, M2.5 x .45 mm Thread
Supplier:  MSC 97643241
Encompass
This stem is designed for use with pocket dial test indicators, providing a reliable and secure attachment point for accurate measurements. With a 3/8 inch diameter and an M2.5 x .45 mm thread, it ensures compatibility and stable performance across various precision testing applications.
  • Accessory type: Stem
  • Compatible with pocket dial test indicators
  • Stem diameter: 3/8 in
  • Thread type: M2.5 x .45 mm
Catalog No. 50-005-30156