Learn More
MSC Mitutoyo Roughness Specimen, 3.00μm Ra, for SJ-400
Supplier: MSC 03300381
This roughness specimen provides a precise 3.00μm Ra reference for calibrating surface roughness measurement instruments. It is specifically designed for use with the SJ-400 and other portable surface roughness testers, ensuring accurate and repeatable measurements.
- Precision roughness specimen
- Features 3.00μm Ra value
- Designed for use with SJ-400
- Compatible with portable surface roughness testers