Learn More
MSC Mitutoyo Surface Roughness Detector, 4 mN, 10 μm/90° Tip, Soft Materials
Supplier:  MSC 03300340
Encompass
The Mitutoyo 178-391 is a surface roughness detector designed for use with Surftest Portable Surface Roughness Testers. It features a 4 mN measuring force, a 10 μm tip radius, and a 90° tip angle, making it suitable for measuring soft materials and small holes.
  • Designed for use with portable surface roughness testers
  • Features a 4 mN measuring force
  • Equipped with a 10 μm tip radius
  • Provides a 90° tip angle
  • Ideal for detecting roughness on soft materials
  • Suitable for measurements in small holes
  • Utilizes a diamond stylus tip for precision
Catalog No. 50-005-29792